Lok-Tight™ Contact Plates, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
P33 P99 P09 P93 P98 P80 P15 P42 P36 P51 P34 P46 P27
CA89407-396PK 204.56 CAD
CA89407-396 CA89407-482 CA89407-510 CA89407-708 CA89407-714 CA89407-728 CA89407-816 CA89407-842 CA89407-860 CA89407-878 CA89407-946 CA89408-026 10153-328 76308-442
Lok-Tight™ Contact Plates, Hardy Diagnostics
Media for Microbiology Prepared Media for Microbiology
An added measure to ensure sterility in your environmental testing program.
  • Friction lid design keeps lids in place
  • Lok-Tight™ optional locking feature means lids stay secured during sampling and transport
  • Each plate has a grid molded into the bottom of the plate to aid in the counting of microorganisms
Hardy Diagnostics contact plates are recommended for use in the cultivation of microorganisms from environmental surfaces.