Brilliant Green Bile Broth, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
K09 K66
CA89407-398PK 453.87 CAD
CA89407-398 CA89407-400
Brilliant Green Bile Broth, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
For the detection of coliforms in water and food, by .