Buffered Peptone Water, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
K107 K195
CA89407-436PK 528.7 CAD
CA89407-436 CA89407-438
Buffered Peptone Water, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
A preenrichment used for increasing the recovery of injured Salmonella species from food and other specimens prior to selective enrichment and isolation.