Violet Red Bile Glucose Agar (VRBGA), CRITERION™, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
C7270 C7271 C7272 C7273
CA89406-242EA 193.91 CAD
CA89406-242 CA89406-244 CA89406-246 CA89406-248
Violet Red Bile Glucose Agar (VRBGA), CRITERION™, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
CRITERION™ Violet Red Bile Glucose Agar is recommended for the detection and enumeration of Enterobacteriaceae in food and dairy products.