Tryptic Soy Broth (TSB), CRITERION™, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
C7140 C7141 C7142 C7143 C7144
CA89406-176EA 198.43 CAD
CA89406-176 CA89406-178 CA89406-180 CA89406-182 CA89406-184
Tryptic Soy Broth (TSB), CRITERION™, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
CRITERION™ Tryptic Soy Broth is recommended for use as a general purpose medium for the isolation and cultivation of a wide variety of microorganisms.
  • Wide mouth opening - reduces dust and allows easy scooping
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers
  • High solubility - dissolves quickly
  • Custom and bulk products available - to your specifications
Certifications: BSE Free