Tergitol 7 Broth, CRITERION™, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
C7910 C7911
CA89406-568EA 156.74 CAD
CA89406-568 CA89406-570
Tergitol 7 Broth, CRITERION™, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
CRITERION™ Tergitol 7 Broth is used for the selective cultivation and differentiation of coliform bacteria
  • Wide mouth opening - reduces dust and allows easy scooping
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers
  • High solubility - dissolves quickly
  • Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container.
Certifications: BSE Free