Sabdex Agar, CRITERION™, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
C6810
C6811
C6812
C6813
CA89405-998EA
180.52
CAD
CA89405-998
CA89406-000
CA89406-002
CA89406-004
Sabdex Agar, CRITERION™, Hardy Diagnostics
Media for Microbiology
Dehydrated Media for Microbiology
CRITERION™ Sabouraud Dextrose Agar (Sabdex) is recommended for the isolation, cultivation, and maintenance of non-pathogenic and pathogenic species of fungi and yeasts
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container.
Certifications: BSE Free