Rappaport-Vassiliadis Broth, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
C8780 C8781 C8782 C8783
CA89406-796EA 211.31 CAD
CA89406-796 CA89406-798 CA89406-800 CA89406-802
Rappaport-Vassiliadis Broth, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
Hardy Diagnostics' CRITERION™ Rappaport-Vassiliadis Broth is recommended for the selective enrichment of Salmonella spp. from food samples.
  • Wide mouth opening - reduces dust and allows easy scooping
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers
  • High solubility - dissolves quickly
  • Custom and bulk products available - to your specifications
CRITERION™ Dehydrated Culture Media by Hardy Diagnostics is formulated to meet or exceed the highest standards. Compare these features…
Hardy Diagnostics' CRITERION™ Rappaport-Vassiliadis Broth is recommended for the selective enrichment of Salmonella spp
from food samples.
Certifications: BSE Free