CRITERION™ Proteose Peptone, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
C6651 C6652 C6653
CA89405-922EA 367.4 CAD
CA89405-922 CA89405-924 CA89405-926
CRITERION™ Proteose Peptone, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
CRITERION™ Proteose Peptone is used as a nitrogen source for use in media for culturing nutritionally fastidious bacteria.
  • Wide mouth opening
  • Reduces dust and allows easy scooping
  • Convenient handgrip
  • 'No slip' finger holds
  • Stackable containers
  • High solubility; dissolves quickly
  • Custom and bulk products available
Custom and bulk products available. BSE Free documentation available. Over 30 years of formulation experience.
Certifications: BSE Free