Phenylethanol Agar (PEA), CRITERION™, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
C6610
C6611
C6612
C6613
CA89405-890EA
212.26
CAD
CA89405-890
CA89405-892
CA89405-894
CA89405-896
Phenylethanol Agar (PEA), CRITERION™, Hardy Diagnostics
Media for Microbiology
Dehydrated Media for Microbiology
CRITERION™ Phenylethanol Agar (PEA) is recommended for use in the cultivation and selective isolation of gram-positive bacteria
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container.
Certifications: BSE Free