CRITERION™ Peptic Digest of Animal Tissue, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
C7480
C7481
C7482
C7483
CA89406-348EA
205.61
CAD
CA89406-348
CA89406-350
CA89406-352
CA89406-354
CRITERION™ Peptic Digest of Animal Tissue, Hardy Diagnostics
Media for Microbiology
Dehydrated Media for Microbiology
CRITERION™ Peptic Digest of Animal Tissue is a source of nutritious peptones and is capable of supporting the growth of a variety of microorganisms
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container. Custom and bulk products available. BSE Free documentation available. Over 30 years of formulation experience.
Certifications: BSE Free