Pancreatic Digest of Gelatin, CRITERION™, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
C6540 C6541 C6542 C6543
CA89405-840EA 174.93 CAD
CA89405-840 CA89405-842 CA89405-844 CA89405-846
Pancreatic Digest of Gelatin, CRITERION™, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
CRITERION™ Pancreatic Digest of Gelatin is a growth promoting peptone used in microbiological culture media, specifically in the cultivation of most nonfastidious microorganisms
  • Wide mouth opening - reduces dust and allows easy scooping
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers
  • High solubility - dissolves quickly
  • Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container.
Certifications: BSE Free