Nutrient Agar with MUG, CRITERION™, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
C7690 C7691 C7692 C7693
CA89406-484EA 169.12 CAD
CA89406-484 CA89406-486 CA89406-488 CA89406-490
Nutrient Agar with MUG, CRITERION™, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
CRITERION™ Nutrient Agar with MUG is primarily used in the detection and enumeration of Escherichia coli from water samples
  • Wide mouth opening - reduces dust and allows easy scooping
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers
  • High solubility - dissolves quickly
  • Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container.
Certifications: BSE Free