MacConkey Agar with Sorbitol, CRITERION™, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
C6170 C6171 C6172 C6173
CA89405-644EA 171.49 CAD
CA89405-644 CA89405-646 CA89405-648 CA89405-650
MacConkey Agar with Sorbitol, CRITERION™, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
CRITERION™ MacConkey Agar with Sorbitol is to be used as a selective and differential medium for the detection of enterohemorrhagic Escherichia coli 0157:H7. Dehydratedculture media with engineered container.
· Wide mouth opening - reduces dust formation by allowing the easy entry of a scoop
· Convenient handgrip - 'no slip' finger holds
· Stackable containers - for efficient storage
· Increased solubility - dissolves quickly
· Large selection, over 200 formulas available
· Custom and bulk products available - to your specifications
· Tested for optimal performance
· BSE Free (contact for specifics)
· Over 30 years of formulation experience
  • Wide mouth opening - reduces dust and allows easy scooping
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers
  • High solubility - dissolves quickly
  • Custom and bulk products available - to your specifications
Custom and bulk products available. BSE Free documentation available. Over 30 years of formulation experience
CRITERION™ MacConkey Agar with Sorbitol is to be used as a selective and differential medium for the detection of enterohemorrhagic Escherichia coli 0157:H7
Dehydratedculture media with engineered container.
Certifications: BSE Free