m PA-C Agar, CRITERION™, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
C7960 C7961 C7962 C7963
CA89406-604EA 193.36 CAD
CA89406-604 CA89406-606 CA89406-608 CA89406-610
m PA-C Agar, CRITERION™, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
CRITERION™ m PA-C Agar is a modified formulation for the cultivation and enumeration of Pseudomonas aeruginosa in water by membrane filtration
  • Wide mouth opening - reduces dust and allows easy scooping
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers
  • High solubility - dissolves quickly
  • Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container.
Certifications: BSE Free