m FC Broth Base, CRITERION™, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
C8220
C8221
C8222
C8223
CA89406-724EA
172.06
CAD
CA89406-724
CA89406-726
CA89406-728
CA89406-730
m FC Broth Base, CRITERION™, Hardy Diagnostics
Media for Microbiology
Dehydrated Media for Microbiology
CRITERION™ m FC Broth Base is recommended for use with added Rosolic Acid for the cultivation and enumeration of fecal coliforms using membrane filtration technique at elevated temperatures
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container.
Certifications: BSE Free