Luria Broth Base, Miller, CRITERION™, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
C6320
C6321
C6322
C6323
CA89405-718EA
187.93
CAD
CA89405-718
CA89405-720
CA89405-722
CA89405-724
Luria Broth Base, Miller, CRITERION™, Hardy Diagnostics
Media for Microbiology
Dehydrated Media for Microbiology
CRITERION™ Luria Broth Base, Miller is used for maintaining and propagating Escherichia coli in molecular microbiology procedures with or without added glucose
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container.
Certifications: BSE/TSE Free