Lactose, (Milk Sugar), CRITERION™, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
C5950
C5951
C5952
C5953
CA89405-536EA
218.74
CAD
CA89405-536
CA89405-538
CA89405-540
CA89405-542
Lactose, (Milk Sugar), CRITERION™, Hardy Diagnostics
Media for Microbiology
Dehydrated Media for Microbiology
CRITERION™ Lactose (Milk Sugar) is chemically pure D-lactose monohydrate
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
It is added to culture media as a source of energy for bacteria and for the determination of fermentation reactions. Dehydratedculture media with engineered container.
Certifications: BSE Free