Dextrose Tryptone Broth, CRITERION™, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
C5631
C5632
C5633
C5634
CA89405-396EA
300.42
CAD
CA89405-396
CA89405-398
CA89405-400
CA89405-402
Dextrose Tryptone Broth, CRITERION™, Hardy Diagnostics
Media for Microbiology
Dehydrated Media for Microbiology
CRITERION™ Dextrose Tryptone Broth for the cultivation of thermophilic “flat sour” microorganisms related to the spoilage of food
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
Dehydrated culture media with engineered container.
Certifications: BSE Free