Dextrose Tryptone Agar, CRITERION™, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
C5620 C5621 C5622 C5623
CA89405-386EA 162.95 CAD
CA89405-386 CA89405-388 CA89405-390 CA89405-392
Dextrose Tryptone Agar, CRITERION™, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
CRITERION™ Dextrose Tryptone Agar is used for cultivating thermophilic "flat-sour" microorganisms associated with food spoilage
  • Wide mouth opening - reduces dust and allows easy scooping
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers
  • High solubility - dissolves quickly
  • Custom and bulk products available - to your specifications
Dehydrated culture media with engineered container.
Certifications: BSE Free