Dextrose Broth, CRITERION™, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
C5600 C5601 C5602 C5603
CA89405-378EA 159.67 CAD
CA89405-378 CA89405-380 CA89405-382 CA89405-384
Dextrose Broth, CRITERION™, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
CRITERION™ Dextrose Broth is recommended for the cultivation of fastidious microorganisms and for the detection of gas from enteric bacilli
  • Wide mouth opening - reduces dust and allows easy scooping
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers
  • High solubility - dissolves quickly
  • Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container.
Certifications: BSE Free