D/E Neutralizing Broth, CRITERION™, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
C7370 C7371 C7372 C7373
CA89406-308EA 174.73 CAD
CA89406-308 CA89406-310 CA89406-312 CA89406-314
D/E Neutralizing Broth, CRITERION™, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
CRITERION™ D/E Neutralizing Broth is an employed for neutralizing of antiseptics and disinfectants and detecting organisms remaining after treatment
  • Wide mouth opening - reduces dust and allows easy scooping
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers
  • High solubility - dissolves quickly
  • Custom and bulk products available - to your specifications
This broth is especially suited for environmental sampling where neutralization of the chemical is important to determine its bactericidal activity. Dehydratedculture media with engineered container.
Certifications: BSE Free