Brain Heart Infusion Broth, CRITERION™, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
C5140 C5141 C5142 C5143 C5144
CA89405-136EA 192.19 CAD
CA89405-136 CA89405-138 CA89405-140 CA89405-142 76308-444
Brain Heart Infusion Broth, CRITERION™, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
CRITERION™ Brain Heart Infusion Broth is a general purpose nutrient medium recommended for the cultivation and isolation of a variety of fastidious and nonfastidious microorganisms, including bacteria, yeasts, and mold
  • Wide mouth opening - reduces dust and allows easy scooping
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers
  • High solubility - dissolves quickly
  • Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container.
Certifications: BSE Free