Bile Esculin Azide Broth, CRITERION™, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
C5190 C5191 C5192 C5193
CA89405-176EA 248.16 CAD
CA89405-176 CA89405-178 CA89405-180 CA89405-182
Bile Esculin Azide Broth, CRITERION™, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
CRITERION™ Bile Esculin Azide Dehydrated Culture Media are recommended for the isolation and differentiation of group D streptococci from non-group D
  • Wide mouth opening - reduces dust and allows easy scooping
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers
  • High solubility - dissolves quickly
  • Custom and bulk products available - to your specifications
CRITERION™ Dehydratedculture media with engineered container.
Certifications: BSE Free