CRITERION™ Agar, Plant Tissue Culture, Grade Purified, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
C8951 C8952 C8953
CA89406-846EA 991.09 CAD
CA89406-846 CA89406-848 CA89406-850
CRITERION™ Agar, Plant Tissue Culture, Grade Purified, Hardy Diagnostics
Cell Culture Media
CRITERION™ Agar, Plant Tissue Culture, Grade Purified, used for culturing plant tissues.
  • Wide mouth opening reduces dust and allows easy scooping
  • Convenient handgrip with 'no slip' finger holds
  • Stackable containers
  • High solubility
  • Custom and bulk products available
Plant tissue culturing is the process of propagating plants in culture media, usually to control the environment in such a way to achieve strong seedlings for mass production.
Certifications: BSE Free