Violet Red Bile Agar, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
G178
G78
U296
U96
CA89407-194PK
185.75
CAD
CA89407-194
CA89407-346
CA89408-164
CA89408-166
Violet Red Bile Agar, Hardy Diagnostics
Media for Microbiology
Dehydrated Media for Microbiology
For the detection and enumeration of coliforms in food, water, and dairy products.