Tryptic Soy Agar (TSA), Soybean Casein Digest Agar
Supplier: HARDY DIAGNOSTICS CA
G151 G41 G60 G60BX G62 J108 Q85 U49 U260 W64 U360 U361 U158 Q58 H19 L60 U412 U174 W41 U373
CA89407-176PK 54 CAD
CA89407-176 CA89407-290 CA89407-330 CA89407-334 CA89407-336 CA89407-390 CA89407-984 CA89408-000 CA89408-002 CA89408-004 CA89408-006 CA89408-008 CA89408-010 CA89408-016 CA89408-020 CA89408-022 CA89408-024 CA89408-032 CA89408-042 CA75994-474
Tryptic Soy Agar (TSA), Soybean Casein Digest Agar
Media for Microbiology Dehydrated Media for Microbiology
For use as a general growth medium for the isolation and cultivation of microorganisms.
  • General growth medium
  • For pure cultures
This medium is also recommended for use in the cultivation, storage, maintenance, and transportation of pure cultures of microorganisms.