StaphTE x ™ Blue, latex Agglutination, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
ST50 ST200 ST1000
CA89407-930EA 594.33 CAD
CA89407-930 CA89407-932 CA89407-934
StaphTE x ™ Blue, latex Agglutination, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
Produces clear and reliable results when testing for Staphylococcus aureus. The plasma coated blue latex beads create the easiest read-out ever! Positives rapidly agglutinate the blue beads on a white background, while negatives remain consistently smooth.
The StaphTEX™ Blue kit tests for both the Clumping Factor and Protein A found in S. aureus. Clumping Factor is detected in both the free and bound form. The easy procedure involves placing several colonies within the raised circle on a white latex agglutination card (PL092HD) sold separately. A drop of the blue latex solution is added and mixed with four inch flat-end mixing sticks (PL091HD) sold separately. Positives begin to clump immediately. The entire procedure takes less than one minute. Liquid controls are included.
Produces clear and reliable results when testing for Staphylococcus aureus
The plasma coated blue latex beads create the easiest read-out ever! Positives rapidly agglutinate the blue beads on a white background, while negatives remain consistently smooth.