Standard Methods Agar (Plate Count Agar), Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
G43 Q21 U395 U295 U95
CA89407-322PK 68.56 CAD
CA89407-322 CA89407-324 CA89407-938 CA89407-940 CA89407-942 CA89407-944
Standard Methods Agar (Plate Count Agar), Hardy Diagnostics
Media for Microbiology Prepared Media for Microbiology
For the cultivation and enumeration of microorganisms in dairy products, food, and water, and the cultivation and differentiation of acid producing bacteria.
Order Now
SPECIFICATIONS
Environmentally Preferable
Storage conditions 2-8°C –2-30°C