Sabdex (Sabouraud Dextrose) Agar
Supplier: HARDY DIAGNOSTICS CA
G159 U227 Q83 U352 U353 H39 U73 K54 W70 L40 W71 U175 U431 Q75 Q31 W74 W73
CA89407-180PK 128.37 CAD
CA89407-180 CA89407-868 CA89407-870 CA89407-872 CA89407-874 CA89407-880 CA89407-882 CA89407-884 CA89407-894 CA89407-896 CA89407-898 CA89407-900 CA89407-902 CA89407-904 CA89407-906 CA89407-908 CA76168-836
Sabdex (Sabouraud Dextrose) Agar
Media for Microbiology Dehydrated Media for Microbiology
Sabouraud Dextrose Agar, Sabouraud Dextrose Broth, and Sabouraud Dextrose Agar, Emmons are recommended for the isolation, cultivation, and maintenance of non-pathogenic and pathogenic species of fungi and yeasts.
Ideal for environmental monitoring in a clean room or clean bench setting.
Sabouraud Dextrose Agar with Chloramphenicol, Sabouraud Dextrose Agar with Chloramphenicol and Gentamicin, and Sabouraud Dextrose Agar with Chloramphenicol and Tetracycline are recommended for the selective isolation of fungi and yeasts from clinical and nonclinical specimens.
Caution: Not intended to be used for the diagnosis of human disease.