Nutrient Agar, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
G114
Q23
W51
L20
CA89407-140PK
186.38
CAD
CA89407-140
CA89407-780
CA89407-782
CA89407-784
Nutrient Agar, Hardy Diagnostics
Media for Microbiology
Dehydrated Media for Microbiology
Nutrient Agar formulations are general purpose growth media recommended for use in the isolation and cultivation of non-fastidious microorganisms
Also, for the detection and enumeration of E. coli in water by membrane filtration.