MR-VP Broth, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
K237 K37
CA89407-756PK 255.12 CAD
CA89407-756 CA89407-758
MR-VP Broth, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
Methyl Red - Voges Proskauer broth for the identification of gram negative bacilli.