Motility Test Medium, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
Q10 Q11
CA89407-752PK 289.3 CAD
CA89407-752 CA89407-754
Motility Test Medium, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
Motility Test Medium with and without Triphenyltetrazolium Chloride (TTC) is recommended for the detection of motility in gram-negative enteric bacilli
TTC is added to enhance visualization of bacterial growth.