m-HPC (Heterotrophic Plate Count) Agar, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
G195 G95
CA89407-212PK 356.22 CAD
CA89407-212 CA89407-694
m-HPC (Heterotrophic Plate Count) Agar, Hardy Diagnostics
Media for Microbiology Prepared Media for Microbiology
For the enumeration of heterotrophic microorganisms in water samples using membrane filtration technique.
Order Now
SPECIFICATIONS
Environmentally Preferable
Storage conditions 2-8°C