MacConkey Agar
Supplier: HARDY DIAGNOSTICS CA
G35 G35BX G36 Q63
CA89407-268PK 57.91 CAD
CA89407-268 CA89407-270 CA89407-272 CA89407-274 CA76006-314
MacConkey Agar
Media for Microbiology Dehydrated Media for Microbiology
For the isolation and differentiation of Gram negative bacteria, especially the Enterobacteriaceae group.