Letheen Broth, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
K118
K106
K207
CA89407-664PK
302.48
CAD
CA89407-664
CA89407-666
CA89407-674
Letheen Broth, Hardy Diagnostics
Media for Microbiology
Dehydrated Media for Microbiology
For use in determining bactericidal activity of quaternary ammonium compounds.