Lauryl Tryptose Broth, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
K33
K32
K61
K338
CA89407-684PK
260.16
CAD
CA89407-684
CA89407-686
CA89407-688
75813-966
Lauryl Tryptose Broth, Hardy Diagnostics
Media for Microbiology
Dehydrated Media for Microbiology
For the detection of coliforms in water and food
Contains a durham tube for gas detection.