Kligler Iron Agar (KIA), Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
L70
R70
CA89407-626PK
573.24
CAD
CA89407-626
CA89407-628
Kligler Iron Agar (KIA), Hardy Diagnostics
Media for Microbiology
Dehydrated Media for Microbiology
For the identification of gram negative enteric bacteria based on hydrogen sulfide production and the fermentation of dextrose and lactose.