Fluid A, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
U19
U109
U349
U249
CA89407-518PK
523.63
CAD
CA89407-518
CA89407-520
CA89407-522
CA89407-530
CA89407-532
Fluid A, Hardy Diagnostics
Media for Microbiology
Dehydrated Media for Microbiology
A rinsing and diluting fluid for use in USP testing protocols
Contains peptone 0.1%.
Multiple bottle types and caps available.