EnviroTrans™ Saline 0.85%, 0.5 ml Swab
Supplier: HARDY DIAGNOSTICS CA
SRK45 SRK55 SRK25 SRK20 SRK05 SRK30 SRK35
CA89407-470PK 93.77 CAD
CA89407-470 CA89407-512 CA89407-672 CA89407-764 CA89407-912 CA89407-914 CA89407-916
EnviroTrans™ Saline 0.85%, 0.5 ml Swab
Media for Microbiology Prepared Media for Microbiology
A ready-to-use swab and tube pre-filled with your choice of five solutions to fit your specific needs! Collect and transport your environmental sample with efficiency and ease
0.85% Saline is recommended for the detection of microorganisms found on surfaces. Osmotically neutral transport solution. Contains no neutralizing agents or growth factors, therefore is recommended for enumeration methods.
Simply remove the cap and attached Dacron® swab, collect the sample, recap, and send to the lab. Each EnviroTrans™ tube contains broth or buffer. The entire unit is terminally sterilized. Excellent for detection or enumeration of microorganisms from environmental surfaces or equipment.
Industry: Breweries, wineries and distilleries, cannabis QA testing, cosmetics, environmental, food safety.