Dilu-Lok™ Dilution Vial with Hinged Cap
Supplier: HARDY DIAGNOSTICS CA
The Dilu-Lok™ eliminates the time and money spent on preparing your dilution vials! Our pre-filled dilution vial is designed especially for the food, dairy, pharmaceutical, and water industries. The vials are filled to 25, 90, or 99 ml with a variety of buffering and enrichment solutions. Each lot is meticulously tested for sterility and fill volume.
- Heavy duty: Vials are made of sturdy polypropylene with a leak-resistant seal that holds up even during air transport
- Leak resistant: Re-engineered for an improved seal, easy-to-remove, perforated, shrink wrap ensures product integrity
- Optically clear: vials are made of clarified polypropylene for clear viewing
Large opening: A wide 45 mm opening accommodates most sample sizes. Each vial is clearly marked with a 90 and 99 ml fill line.
One-handed: Flip-top lid allows for easy one-handed opening and closing to make pipetting easier. Saves unnecessary motion! No awkward screw caps or plastic strips to tear off.
Variety: Available with peptone water, buffered peptone water, Butterfield's buffer, deionized water, phosphate buffer with MgCl, and an empty sterile container.
Industry: Breweries, wineries and distilleries, canabis QA testing, cosmetics, environmental, food safety.
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...