Butterfield's Phosphate Buffer, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
U150
K109
K208
K119
U190
K209
U290
CA89407-406PK
1359.38
CAD
CA89407-406
CA89407-408
CA89407-410
CA89407-412
CA89407-414
CA89407-416
CA89407-440
Butterfield's Phosphate Buffer, Hardy Diagnostics
Media for Microbiology
Dehydrated Media for Microbiology
For the preparation of dilutions for plate count and other laboratory tests, pH 7.2.