XLD Agar, CRITERION™ Dehydrated Culture Media, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
C7320 C7321 C7322
CA89406-274EA 234.76 CAD
CA89406-274 CA89406-276 CA89406-278
XLD Agar, CRITERION™ Dehydrated Culture Media, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
CRITERION™ XLD Agar is recommended for use as a selective and differential medium for the isolation of gram-negative enteric pathogens such as Salmonella and Shigella from fecal and other specimens. Dehydratedculture media with engineered container.
  • Wide mouth opening - reduces dust and allows easy scooping
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers
  • High solubility - dissolves quickly
  • Custom and bulk products available - to your specifications
Custom and bulk products available. BSE Free documentation available. Over 30 years of formulation experience
CRITERION™ XLD Agar is recommended for use as a selective and differential medium for the isolation of gram-negative enteric pathogens such as Salmonella and Shigella from fecal and other specimens
Dehydratedculture media with engineered container.
Certifications: BSE Free