Brucella Agar, CRITERION™, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
C5300 C5301 C5302 C5303
CA89405-232EA 159.1 CAD
CA89405-232 CA89405-234 CA89405-236 CA89405-238
Brucella Agar, CRITERION™, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
CRITERION™ Brucella Agar is used for the cultivation of Brucella species, and other fastidious organisms
  • Wide mouth opening - reduces dust and allows easy scooping
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers
  • High solubility - dissolves quickly
  • Custom and bulk products available - to your specifications
CRITERION™ Dehydratedculture media with engineered container.
Certifications: BSE Free