Ampicillin Dextrin Agar Base, CRITERION™, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
C7580 C7581
CA89406-412EA 229.88 CAD
CA89406-412 CA89406-414 CA89406-416 CA89406-418
Ampicillin Dextrin Agar Base, CRITERION™, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
CRITERION™ Pharmaceutical Grade Agar is primarily a solidifying agent which is purified to remove extraneous matter, pigments and salts
  • Wide mouth opening - reduces dust and allows easy scooping
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers
  • High solubility - dissolves quickly
  • Custom and bulk products available - to your specifications
Pharmaceutical grade is suitable for use in preparing culture media (requires Ampicillin Selective Supplement SR136E)
Certifications: BSE Free