CRITERION™ Yersinia Selective Agar Base, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
C5390
C5391
CA89405-282EA
202.76
CAD
CA89405-282
CA89405-284
CA89405-286
CA89405-288
CRITERION™ Yersinia Selective Agar Base, Hardy Diagnostics
Media for Microbiology
Dehydrated Media for Microbiology
CRITERION™ Yersinia Selective Agar (CIN) Base is recommended for use in the isolation and differentiation of Yersinia and Aeromonas species from clinical specimens, environmental samples, and food sources
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
Yersinia Antimicrobial Supplement CN, product #231961, is required for this medium.
Certifications: BSE Free