CRITERION™ Yeast Extract, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
C7340 C7341 C7342 C7343 C7344
CA89406-290EA 173.61 CAD
CA89406-290 CA89406-292 CA89406-294 CA89406-296 CA89406-298
CRITERION™ Yeast Extract, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
CRITERION™ Yeast Extract is prepared and standardized for bacteriological use
  • Wide mouth opening - reduces dust and allows easy scooping
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers
  • High solubility - dissolves quickly
  • Custom and bulk products available - to your specifications
It is an excellent stimulator of bacterial growth and is used in culture media in place of, or in addition to, beef extract.
Certifications: BSE Free