W-L Differential Medium, CRITERION™, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
C7290 C7291 C7292 C7293
CA89406-258EA 204.85 CAD
CA89406-258 CA89406-260 CA89406-262 CA89406-264
W-L Differential Medium, CRITERION™, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
CRITERION™ W-L Differential Medium is primarily for use in the isolation of bacteria encountered in brewing and industrial fermentation processes.
Dehydratedculture media with engineered container.
  • Wide mouth opening - reduces dust and allows easy scooping
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers
  • High solubility - dissolves quickly
  • Custom and bulk products available - to your specifications
Custom and bulk products available. BSE Free documentation available. Over 30 years of formulation experience
CRITERION™ W-L Differential Medium is primarily for use in the isolation of bacteria encountered in brewing and industrial fermentation processes.
Certifications: BSE Free