Vogel and Johnson Agar, CRITERION™, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
C7281
C7282
CA89406-252EA
1032.2
CAD
CA89406-252
CA89406-254
Vogel and Johnson Agar, CRITERION™, Hardy Diagnostics
Media for Microbiology
Dehydrated Media for Microbiology
CRITERION™ Vogel and Johnson Agar is a selective agar medium for the detection of coagulase-positive, mannitol-positive Staphylococcus aureus strains in clinical specimens and foods
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
Potassium Tellurite, product #SR030J, is required for this medium.
Certifications: BSE Free