Violet Red Bile Agar with MUG, CRITERION™, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
CRITERION™ Violet Red Bile Agar with MUG is primarily for use in the detection of Escherichia coli and total coliforms in food and dairy products.
CRITERION™ Dehydrated Culture Media and container.
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
Custom and bulk products available. BSE Free documentation available. Over 30 years of formulation experience
CRITERION™ Violet Red Bile Agar with MUG is primarily for use in the detection of Escherichia coli and total coliforms in food and dairy products.
Certifications: BSE Free